Learn More
Medchemexpress LLC 9,10-Anthracenedione, 3-hydroxy-1-methoxy-2-methyl- | 7460-43-7 | 268.26 | 1 ML
SDP
Catalog No. 5000416180
Encompass_Preferred
Rubiadin-1-methyl ether is a natural anthraquinone isolated from *Morinda officinalis How*. It inhibits osteoclastic bone resorption by blocking the phosphorylation of NF-κB p65 and the degradation of IκBα, as well as decreasing the nuclear translocation of p65. It is for research use only and not sold to patients.
  • Natural anthraquinone isolated from *Morinda officinalis How*.
  • Inhibits osteoclastic bone resorption.
  • Acts by inhibiting the phosphorylation of NF-κB p65 and the degradation of IκBα.
  • Decreases the nuclear translocation of p65.
  • Purity: 98.57%.
Catalog No. 50-004-16180 Supplier Medchemexpress LLC Supplier No. HYN195610MM/1ML
Rubiadin-1-methyl ether is a natural anthraquinone isolated from *Morinda officinalis How*. It inhibits osteoclastic bone resorption by blocking the phosphorylation of NF-κB p65 and the degradation of IκBα, as well as decreasing the nuclear translocation of p65. It is for research use only and not sold to patients.
  • Natural anthraquinone isolated from *Morinda officinalis How*.
  • Inhibits osteoclastic bone resorption.
  • Acts by inhibiting the phosphorylation of NF-κB p65 and the degradation of IκBα.
  • Decreases the nuclear translocation of p65.
  • Purity: 98.57%.