Learn More
AdipoGen 9 10-Anthracenediyl-bismethyl
SDP
Catalog No. 502561808
Encompass_Preferred
Chemical. CAS 307554-62-7. Formula C22H18O8. MW 410.37. Synthetic. Reagent for the assay of O2 with better characteristics than 9,10-anthracenediyl-bis-dipropionic acid.
Catalog No. 50-256-1808 Supplier AdipoGen Supplier No. CDXA0183M200
Chemical. CAS 307554-62-7. Formula C22H18O8. MW 410.37. Synthetic. Reagent for the assay of O2 with better characteristics than 9,10-anthracenediyl-bis-dipropionic acid.