Learn More
AdipoGen 9,10-Bis(chloromethyl)anthracene (5 g)
SDP
Catalog No. 502054295
Encompass_Preferred
Chemical. CAS: 10387-13-0. Formula: C16H12CI2. MW: 275.18. Building block and intermediate for synthesis of fluorescent and luminescent agents. This compound has been used as a cross-linking agent to incorporate 9,10-anthracenylidene (AN) chromophores into linear polystyrene chains.
Catalog No. 50-205-4295 Supplier AdipoGen Supplier No. CDXB0132G005
Chemical. CAS: 10387-13-0. Formula: C16H12CI2. MW: 275.18. Building block and intermediate for synthesis of fluorescent and luminescent agents. This compound has been used as a cross-linking agent to incorporate 9,10-anthracenylidene (AN) chromophores into linear polystyrene chains.