Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 9β,10α-Cholesta-5,7-dien-3β-ol | 5226-01-7 | 99.5% | 384.64 | 10 MG
SDP

Catalog No. 5000438949
Encompass_Preferred

Lumisterol 3 (9β,10α-Cholesta-5,7-dien-3β-ol) is a normal human secosteroid metabolite, classified as a vitamin D3 photoisomer derivative. It is used in the preparation of vitamin D.

  • Source: endogenous metabolite, human gut microbiota metabolites
  • Appearance: solid, off-white to light yellow
  • Molecular formula: C27H44O
  • Structure classification: steroids
  • Shipping: shipped with dry ice
  • Storage: -80°C, protected from light, under nitrogen. Solutions are unstable, freshly prepared recommended.
  • Biological activity: reduces UVB-induced cellular oxidant formation, inhibits proliferation of UVB-irradiated keratinocytes, promotes cellular DNA damage repair, increases nuclear/cytosolic Nrf2 ratio and nuclear p53 expression, enhances Nrf2-regulated antioxidant proteins.

Catalog No. 50-004-38949 Supplier Medchemexpress LLC Supplier No. HY11202310MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Lumisterol 3 (9β,10α-Cholesta-5,7-dien-3β-ol) is a normal human secosteroid metabolite, classified as a vitamin D3 photoisomer derivative. It is used in the preparation of vitamin D.

  • Source: endogenous metabolite, human gut microbiota metabolites
  • Appearance: solid, off-white to light yellow
  • Molecular formula: C27H44O
  • Structure classification: steroids
  • Shipping: shipped with dry ice
  • Storage: -80°C, protected from light, under nitrogen. Solutions are unstable, freshly prepared recommended.
  • Biological activity: reduces UVB-induced cellular oxidant formation, inhibits proliferation of UVB-irradiated keratinocytes, promotes cellular DNA damage repair, increases nuclear/cytosolic Nrf2 ratio and nuclear p53 expression, enhances Nrf2-regulated antioxidant proteins.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.