Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9,10-Di(2-naphthayl)anthracene (purified by sublimation) 99.0+%, TCI America™
SDP

Catalog No. D5066200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D5066200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D5066200MG Supplier TCI America Supplier No. D5066200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 122648-99-1
Molecular Formula C34H22
Molecular Weight (g/mol) 430.55
MDL Number MFCD00028944
InChI Key VIZUPBYFLORCRA-UHFFFAOYSA-N
PubChem CID 21881242
IUPAC Name 9,10-dinaphthalen-2-ylanthracene
SMILES C1=CC=C2C=C(C=CC2=C1)C3=C4C=CC=CC4=C(C5=CC=CC=C53)C6=CC7=CC=CC=C7C=C6

Specifications

Melting Point 357°C
Color White-Yellow
Quantity 200 mg
Formula Weight 430.55
Percent Purity ≥99.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 9,10-Di(2-naphthayl)anthracene (purified by sublimation)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.