Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9,10-Diethoxyanthracene, 98%

Catalog No. AAA1670514
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1670514 25 g
AAA1670503 1 g
AAA1670506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1670514 Supplier Thermo Scientific Chemicals Supplier No. A1670514
  • $632.50
    $534.65 / Each of 1

    Save $97.85

    15% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 68818-86-0 | C18H18O2 | 266.34 g/mol

9,10-Diethoxyanthracene acts as a photo sensitizing agent for diaryl iodonium salts. It is also used as a sensitizing dye. It is utilized for the preparation of polymetallic complexes through selective pi-coordination.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
9,10-Diethoxyanthracene acts as a photo sensitizing agent for diaryl iodonium salts. It is also used as a sensitizing dye. It is utilized for the preparation of polymetallic complexes through selective pi-coordination.

Solubility
Soluble in toluene.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68818-86-0
Molecular Formula C18H18O2
Molecular Weight (g/mol) 266.34
MDL Number MFCD00209643
InChI Key GJNKQJAJXSUJBO-UHFFFAOYSA-N
Synonym anthracene, 9,10-diethoxy, acmc-1bigo
PubChem CID 111623
IUPAC Name 9,10-diethoxyanthracene
SMILES CCOC1=C2C=CC=CC2=C(C3=CC=CC=C31)OCC

Specifications

Melting Point 149°C to 151°C
Quantity 25 g
Solubility Information Soluble in toluene.
Formula Weight 266.33
Percent Purity 98%
Chemical Name or Material 9,10-Diethoxyanthracene
EINECSNumber 272-401-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.