Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9,10-Difluoro-2,3-dihydro-3-methyl-7-oxo-7H-pyrido[1,2,3-de]-1,4-benzoxazine-6-carboxylic Acid 98.0+%, TCI America™
SDP

Catalog No. O040425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O040425G 25 g
O04045G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O040425G Supplier TCI America Supplier No. O040425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 82419-35-0
Molecular Formula C13H9F2NO4
Molecular Weight (g/mol) 281.215
MDL Number MFCD00226106
InChI Key NVKWWNNJFKZNJO-UHFFFAOYSA-N
Synonym 9,10-difluoro-2,3-dihydro-3-methyl-7-oxo-7h-pyrido 1,2,3-de-1,4-benzoxazine-6-carboxylic acid, ofloxacin q acid, oxygen-fluorine acid, 9,10-difluoro-3-methyl-7-oxo-2,3-dihydro-7h-1,4 oxazino 2,3,4-ij quinoline-6-carboxylic acid, s---9,10-difluoro-2,3-dihydro-3-methyl-7-oxo-7h-pyrido-1,2,3-de-1,4-benzoxazine-6-carboxylic acid, 9,10-difluoro-2,3-dihydro-3-methyl-7-oxo-7h-pyrido 1,2,3-de-1,4-benzoxazine-6-carboxyllic acid, 9,10-difluoro-3-methyl-7-oxo-2,3-dihydro-7h-pyrido 1,2,3-de 1,4 benzoxazine-6-carboxylic acid, levofloxacin related compound b, ofloxacin q acid, +/-, 6,7-difluoro-2-methyl-10-oxo-4-oxa-1-azatricyclo 7.3.1.0?, 1 3 trideca-5 13 ,6,8,11-tetraene-11-carboxylic acid
PubChem CID 534278
SMILES CC1COC2=C3N1C=C(C(=O)C3=CC(=C2F)F)C(=O)O

Specifications

Melting Point 311°C
Color White-Yellow
Quantity 25 g
Formula Weight 281.21
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 9,10-Difluoro-2,3-dihydro-3-methyl-7-oxo-7H-pyrido[1,2,3-de]-1,4-benzoxazine-6-carboxylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.