Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9,10-Dihydrophenanthrene, 95%, Thermo Scientific™

Catalog No. AC114770100
Change view
Click to view available options
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC114770100 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC114770100 Supplier Thermo Scientific Chemicals Supplier No. 114770100

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 776-35-2
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Packaging Glass bottle
Molecular Formula C14H12
Quantity 10g
Beilstein 05, 642
Synonym phenanthrene, 9,10-dihydro, unii-brm9tu2f34, 9,10-dihydro-phenanthrene, dihydrophenanthrene, brm9tu2f34, acmc-1ba4q, phenanthrene,9,10-dihydro, 9,10-dihydrophenanthrene, phenanthrene, 9,10-dihydro-8ci 9ci
InChI Key XXPBFNVKTVJZKF-UHFFFAOYSA-N
SMILES C1CC2=CC=CC=C2C3=CC=CC=C31
IUPAC Name 9,10-dihydrophenanthrene
Molecular Weight (g/mol) 180.25
PubChem CID 13058
Formula Weight 180.25
Chemical Name or Material 9, 10-Dihydrophenanthrene, 95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.