Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Echelon Biosciences Research Labs 9(10)-Nitrooleic Acid, 1 mg, C18H33NO4, 327.24 g/mol, CAS: 875685-44-4
SDP

Catalog No. NC3566296
Encompass

9(10)-Nitrooleic Acid (OA-NO2) is a key lipid signaling molecule that emerges through the nitration of oleic acid during inflammatory conditions. This compound is critical for researchers focused on cellular signaling pathways and inflammation responses. It activates receptors such as TRPA1 and PPARγ and exists as a mixture of 9-nitro and 10-nitro isomers, both produced in equal amounts in vivo.

  • Molecular formula: C18H33NO4
  • Molecular weight: 327.24 g/mol
  • CAS number: 875685-44-4
  • Quantity: 1 mg
  • Storage conditions: -20 °C or below
  • Shipping temperature: Gel ice (primary), cool pack (secondary)
  • Solubility: Methanol, ethanol
  • Intended use: Research and development

Catalog No. NC3566296 Supplier Echelon Biosciences Research Labs Supplier No. L0112
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

9(10)-Nitrooleic Acid (OA-NO2) is a key lipid signaling molecule that emerges through the nitration of oleic acid during inflammatory conditions. This compound is critical for researchers focused on cellular signaling pathways and inflammation responses. It activates receptors such as TRPA1 and PPARγ and exists as a mixture of 9-nitro and 10-nitro isomers, both produced in equal amounts in vivo.

  • Molecular formula: C18H33NO4
  • Molecular weight: 327.24 g/mol
  • CAS number: 875685-44-4
  • Quantity: 1 mg
  • Storage conditions: -20 °C or below
  • Shipping temperature: Gel ice (primary), cool pack (secondary)
  • Solubility: Methanol, ethanol
  • Intended use: Research and development

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.