Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 9,10-phenanthrenedione | 84-11-7 | MFCD00001163 | 99.7% | 208.21 g/mol | C14H8O2 | 100 G
SDP

Catalog No. 5000303306
Encompass_Preferred

9,10-Phenanthrenequinone (9,10-phenanthrenedione; CAS 84-11-7) is a polycyclic quinone used in biochemical research as a claudin-5 (CLDN5) inhibitor. It induces apoptosis via nitric oxide synthase/ROS-dependent pathways and can disrupt endothelial barrier integrity.

  • Molecular formula C14H8O2; molecular weight 208.21 g/mol.
  • High purity: 99.7% (typical; certificate of analysis available).
  • Physical form: solid; available in 10 g, 25 g, and 100 g pack sizes.
  • Application: biochemical probe for claudin-5 inhibition and apoptosis studies.
  • Documentation: certificate of analysis and batch purity data available.

Catalog No. 50-003-03306 Supplier Medchemexpress LLC Supplier No. HYW014507100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

9,10-Phenanthrenequinone (9,10-phenanthrenedione; CAS 84-11-7) is a polycyclic quinone used in biochemical research as a claudin-5 (CLDN5) inhibitor. It induces apoptosis via nitric oxide synthase/ROS-dependent pathways and can disrupt endothelial barrier integrity.

  • Molecular formula C14H8O2; molecular weight 208.21 g/mol.
  • High purity: 99.7% (typical; certificate of analysis available).
  • Physical form: solid; available in 10 g, 25 g, and 100 g pack sizes.
  • Application: biochemical probe for claudin-5 inhibition and apoptosis studies.
  • Documentation: certificate of analysis and batch purity data available.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.