Learn More
Cayman Chemical 9 10a-ntI-6a 7-THCexemp 100ug
SDP
Catalog No. 502945058
Encompass_Preferred
An analytical reference standard that is structurally similar to known phytocannabinoids and is a mixture of (9R,10aS)-anti-Δ6a,7-THC and (9S,10aR)-anti-Δ6a,7-THC enantiomers; provided as an exempt preparation; intended for research and forensic applications
Catalog No. 50-294-5058 Supplier Cayman Chemical Supplier No. 41297100 UG
An analytical reference standard that is structurally similar to known phytocannabinoids and is a mixture of (9R,10aS)-anti-Δ6a,7-THC and (9S,10aR)-anti-Δ6a,7-THC enantiomers; provided as an exempt preparation; intended for research and forensic applications