Learn More
Cayman Chemical 9 10DiHOMEd4 GC MS Standards
SDP
Supplier:  Cayman Chemical 1000999325 UG
Encompass_Preferred
An internal standard for the quantification of 9(10)-DiHOME by GC- or LC-MS
Catalog No. 50-288-9184