Learn More
Cayman Chemical 9 10DiHOMEd4 GC MS Standards
SDP
Catalog No. 502889185
Encompass_Preferred
An internal standard for the quantification of 9(10)-DiHOME by GC- or LC-MS
Catalog No. 50-288-9185 Supplier Cayman Chemical Supplier No. 1000999350 UG
Add to Cart
Add to Cart
An internal standard for the quantification of 9(10)-DiHOME by GC- or LC-MS