Learn More
Cayman Chemical 9,10-Dimethoxypterocarpan-3-O-β-D-glucoside, Purity: ≥98%, CAS: 94367-42-7, 10 mg
SDP
Catalog No. 502911870
Encompass_Preferred
A flavonoid glycoside with anti-inflammatory activity; inhibits LPS-induced NO production in RAW 264.7 macrophages (EC50 = 14.7 µM)
Catalog No. 50-291-1870 Supplier Cayman Chemical Supplier No. 3619210 MG
A flavonoid glycoside with anti-inflammatory activity; inhibits LPS-induced NO production in RAW 264.7 macrophages (EC50 = 14.7 µM)