Learn More
Cayman Chemical 9 10EpOMEd4 GC MS Standards
SDP
Catalog No. 502889198
Encompass_Preferred
An internal standard for the quantification of latanoprost by GC- or LC-mass spectrometry (MS).
Catalog No. 50-288-9198 Supplier Cayman Chemical Supplier No. 1000999525 UG
An internal standard for the quantification of latanoprost by GC- or LC-mass spectrometry (MS).