Learn More
Cayman Chemical 9 10EpOMEd4 GC MS Standards
SDP
Supplier:  Cayman Chemical 1000999550 UG
Encompass_Preferred
An internal standard for the quantification of latanoprost by GC- or LC-mass spectrometry (MS).
Catalog No. 50-288-8462