Learn More
Medchemexpress LLC 9(10H)-acridinone, 10-(3-aminopropyl)-3,4-dimethyl-, (2Z)-2-butenedioate (1:1) | 1204480-26-1 | 99.4% | 396.44 | 25 MG
SDP
Catalog No. 5000347305
Encompass_Preferred
ER-27319 (maleate), an acridone derivative, is a potent and selective SYK inhibitor. It inhibits the tyrosine phosphorylation of SYK and its activity. This compound also inhibits the release of antigen-induced allergic mediators from human and rat mast cells with an IC50 of 10 μM, making it useful for studying allergic diseases.
  • Potent and selective SYK inhibitor.
  • Inhibits tyrosine phosphorylation of SYK and its activity.
  • Inhibits the release of antigen-induced allergic mediators from human and rat mast cells with an IC50 of 10 μM.
  • Useful for study in allergic diseases.
  • Available in solid form, yellow to orange color.
Catalog No. 50-003-47305 Supplier Medchemexpress LLC Supplier No. 25MGHY108489
ER-27319 (maleate), an acridone derivative, is a potent and selective SYK inhibitor. It inhibits the tyrosine phosphorylation of SYK and its activity. This compound also inhibits the release of antigen-induced allergic mediators from human and rat mast cells with an IC50 of 10 μM, making it useful for studying allergic diseases.
  • Potent and selective SYK inhibitor.
  • Inhibits tyrosine phosphorylation of SYK and its activity.
  • Inhibits the release of antigen-induced allergic mediators from human and rat mast cells with an IC50 of 10 μM.
  • Useful for study in allergic diseases.
  • Available in solid form, yellow to orange color.