Learn More
Cayman Chemical 9,10-Anthracenediyl-bis(methylene)dimalonic Acid, Purity: ≥90%, CAS: 307554-62-7, 50 mg
SDP
Catalog No. 502984458
Encompass_Preferred
A reagent used to detect singlet oxygen generation; can be photobleached by singlet oxygen to its corresponding endoperoxide
Catalog No. 50-298-4458 Supplier Cayman Chemical Supplier No. 1958050 MG
A reagent used to detect singlet oxygen generation; can be photobleached by singlet oxygen to its corresponding endoperoxide