Learn More
Cayman Chemical 9 10nthrACndIylbIs methy 250mg
SDP
Catalog No. 502983687
Encompass_Preferred
A reagent used to detect singlet oxygen generation; can be photobleached by singlet oxygen to its corresponding endoperoxide
Catalog No. 50-298-3687 Supplier Cayman Chemical Supplier No. 19580250 MG
A reagent used to detect singlet oxygen generation; can be photobleached by singlet oxygen to its corresponding endoperoxide