Learn More
Medchemexpress LLC 9,12-Octadecadienoic acid (9Z,12Z)-, 1,1',1''-(1,2,3-propanetriyl) ester | 537-40-6 | ≥98.0% | 100 MG
SDP
Catalog No. 5000339110
Encompass_Preferred
Trilinolein is an orally active triglyceride that inhibits the PI3K/Akt, Ras/MEK/ERK signaling pathways, and MMP-2. It can reduce oxidative stress, induce apoptosis, and inhibit cell migration. Trilinolein can be used in the research fields of cardiovascular disease, cerebrovascular disease (such as cerebral ischemia), and non-small cell lung cancer.
  • Inhibits PI3K/Akt, Ras/MEK/ERK signaling pathways, and MMP-2.
  • Reduces oxidative stress.
  • Induces apoptosis.
  • Inhibits cell migration.
  • Useful for research in cardiovascular disease.
  • Useful for research in cerebrovascular disease, such as cerebral ischemia.
  • Useful for research in non-small cell lung cancer.
Catalog No. 50-003-39110 Supplier Medchemexpress LLC Supplier No. HY128393100MG
Trilinolein is an orally active triglyceride that inhibits the PI3K/Akt, Ras/MEK/ERK signaling pathways, and MMP-2. It can reduce oxidative stress, induce apoptosis, and inhibit cell migration. Trilinolein can be used in the research fields of cardiovascular disease, cerebrovascular disease (such as cerebral ischemia), and non-small cell lung cancer.
  • Inhibits PI3K/Akt, Ras/MEK/ERK signaling pathways, and MMP-2.
  • Reduces oxidative stress.
  • Induces apoptosis.
  • Inhibits cell migration.
  • Useful for research in cardiovascular disease.
  • Useful for research in cerebrovascular disease, such as cerebral ischemia.
  • Useful for research in non-small cell lung cancer.