Learn More
Medchemexpress LLC 9,12-Octadecadienoic acid (9Z,12Z)-, 2,3-dihydroxypropyl ester | 2277-28-3 | 98.0% | 354.52 | 1 MG
SDP
Catalog No. 5000343781
Encompass_Preferred
1-Linoleoyl glycerol is a LpPLA2 inhibitor. It acts as a precursor for synthesizing various functional lipids, such as phospholipids. It mitigates inflammation induced by Apolipoprotein CIII (reduction of IL-6).
  • Acts as a LpPLA2 inhibitor
  • Precursor for synthesizing various functional lipids, such as phospholipids
  • Mitigates inflammation induced by Apolipoprotein CIII (reduction of IL-6)
  • (R)-1-Linoleoyl glycerol and (S)-1-Linoleoyl glycerol exhibit Lp-PLA2 inhibitory activities
  • Metabolized by human eosinophils to 13-HODE-G and 13-HODE
  • Metabolized by human neutrophils to produce 15-LO metabolite
Catalog No. 50-003-43781 Supplier Medchemexpress LLC Supplier No. HY11134610MM/1ML
1-Linoleoyl glycerol is a LpPLA2 inhibitor. It acts as a precursor for synthesizing various functional lipids, such as phospholipids. It mitigates inflammation induced by Apolipoprotein CIII (reduction of IL-6).
  • Acts as a LpPLA2 inhibitor
  • Precursor for synthesizing various functional lipids, such as phospholipids
  • Mitigates inflammation induced by Apolipoprotein CIII (reduction of IL-6)
  • (R)-1-Linoleoyl glycerol and (S)-1-Linoleoyl glycerol exhibit Lp-PLA2 inhibitory activities
  • Metabolized by human eosinophils to 13-HODE-G and 13-HODE
  • Metabolized by human neutrophils to produce 15-LO metabolite