Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 9,12-Octadecadienoic acid (9Z,12Z)-, 2-hydroxy-1-(hydroxymethyl)ethyl ester | 3443-82-1 | 99.9% | 500 UG
SDP

Catalog No. 5000340942
Encompass_Preferred

2-Linoleoyl glycerol is a monoacylglycerol that acts as an antagonist and partial agonist at the type 1 cannabinoid CB1 receptor. Its potency can be increased by JZL195, an inhibitor of FAAH and MAGL, and decreased by the CB1 antagonist AM251 and Cannabidiol. As a CB1 antagonist, 2-Linoleoyl glycerol does not enhance, but only attenuates, the activity of the CB1/CB2 receptor ligands such as cannabinoids (AEA) and 2-arachidonoylglycerol (2-AG). It is for research use only.

  • Acts as an antagonist and partial agonist at the type 1 cannabinoid CB1 receptor
  • Potency increased by JZL195, an inhibitor of FAAH and MAGL
  • Activity decreased by the CB1 antagonist AM251 and Cannabidiol
  • Attenuates the activity of CB1/CB2 receptor ligands
  • Molecular weight: 354.52
  • Formula: C21H38O4
  • Appearance: Liquid, colorless to light yellow
  • Storage: Solution, -20°C, 2 years

Catalog No. 50-003-40942 Supplier Medchemexpress LLC Supplier No. HY130311500UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Linoleoyl glycerol is a monoacylglycerol that acts as an antagonist and partial agonist at the type 1 cannabinoid CB1 receptor. Its potency can be increased by JZL195, an inhibitor of FAAH and MAGL, and decreased by the CB1 antagonist AM251 and Cannabidiol. As a CB1 antagonist, 2-Linoleoyl glycerol does not enhance, but only attenuates, the activity of the CB1/CB2 receptor ligands such as cannabinoids (AEA) and 2-arachidonoylglycerol (2-AG). It is for research use only.

  • Acts as an antagonist and partial agonist at the type 1 cannabinoid CB1 receptor
  • Potency increased by JZL195, an inhibitor of FAAH and MAGL
  • Activity decreased by the CB1 antagonist AM251 and Cannabidiol
  • Attenuates the activity of CB1/CB2 receptor ligands
  • Molecular weight: 354.52
  • Formula: C21H38O4
  • Appearance: Liquid, colorless to light yellow
  • Storage: Solution, -20°C, 2 years

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.