Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 9,12-Octadecadienoic acid (9Z,12Z)- | 60-33-3 | 280.45 | 25 MG
SDP

Catalog No. 5000390342
Encompass_Preferred

Linoleic acid is a common polyunsaturated fatty acid (PUFA) found in plant-based oils, nuts, and seeds. It functions as a structural component of membrane phospholipids, maintaining a certain level of membrane fluidity in the transdermal water barrier of the epidermis. This product is suitable for research and analytical applications.

  • Common polyunsaturated fatty acid.
  • Found in plant-based oils, nuts, and seeds.
  • Structural component of membrane phospholipids.
  • Maintains membrane fluidity.
  • Purity (HPLC) is 98.12%.
  • Appearance: colorless to light yellow liquid.

Catalog No. 50-003-90342 Supplier Medchemexpress LLC Supplier No. 25MGHYN0729R
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Linoleic acid is a common polyunsaturated fatty acid (PUFA) found in plant-based oils, nuts, and seeds. It functions as a structural component of membrane phospholipids, maintaining a certain level of membrane fluidity in the transdermal water barrier of the epidermis. This product is suitable for research and analytical applications.

  • Common polyunsaturated fatty acid.
  • Found in plant-based oils, nuts, and seeds.
  • Structural component of membrane phospholipids.
  • Maintains membrane fluidity.
  • Purity (HPLC) is 98.12%.
  • Appearance: colorless to light yellow liquid.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.