Learn More
Medchemexpress LLC 9,12-Octadecadienoic acid (9Z,12Z)- | 60-33-3 | MFCD00064241 | 280.45 | 5 G
SDP
Catalog No. 5000417133
Encompass_Preferred
Linoleic acid is a common polyunsaturated fatty acid (PUFA) found in plant-based oils, nuts, and seeds. It is a structural component of membrane phospholipids, maintaining membrane fluidity of the transdermal water barrier of the epidermis. This product is suitable for research applications.
  • Common polyunsaturated fatty acid
  • Maintains membrane fluidity
  • Purity of 99.92%
  • Appears as a colorless to light yellow liquid
Catalog No. 50-004-17133 Supplier Medchemexpress LLC Supplier No. HYN07295G
Linoleic acid is a common polyunsaturated fatty acid (PUFA) found in plant-based oils, nuts, and seeds. It is a structural component of membrane phospholipids, maintaining membrane fluidity of the transdermal water barrier of the epidermis. This product is suitable for research applications.
  • Common polyunsaturated fatty acid
  • Maintains membrane fluidity
  • Purity of 99.92%
  • Appears as a colorless to light yellow liquid