Learn More
Medchemexpress LLC 9α,15S-dihydroxy-11-oxo-prosta-5Z,13E-dien-1-oic-3,3,4,4-d4 acid | 211105-29-2 | 98.8% | 356.49 g/mol | C20H28D4O5 | 25 UG
SDP
Catalog No. 5000248702
Encompass_Preferred
Prostaglandin D2-d4 is a deuterium-labeled analogue of prostaglandin D2 intended for use as an internal standard or tracer in quantitative analysis. It is suitable for NMR, GC-MS, and LC-MS workflows, and leverages isotopic labeling to provide mass discrimination for more accurate measurements of endogenous or spiked prostaglandin D2.
  • Deuterium-labeled internal standard for accurate quantification.
  • Suitable for NMR, GC-MS, and LC-MS assays.
  • High purity (98.8%).
  • Provided as a solution in methyl acetate for easy use.
  • Stable when stored at -20°C, typical shelf life 2 years.
  • Available in small, trace-analysis pack sizes (e.g., 25 μg).
Catalog No. 50-002-48702 Supplier Medchemexpress LLC Supplier No. HY101988S25UG
Prostaglandin D2-d4 is a deuterium-labeled analogue of prostaglandin D2 intended for use as an internal standard or tracer in quantitative analysis. It is suitable for NMR, GC-MS, and LC-MS workflows, and leverages isotopic labeling to provide mass discrimination for more accurate measurements of endogenous or spiked prostaglandin D2.
  • Deuterium-labeled internal standard for accurate quantification.
  • Suitable for NMR, GC-MS, and LC-MS assays.
  • High purity (98.8%).
  • Provided as a solution in methyl acetate for easy use.
  • Stable when stored at -20°C, typical shelf life 2 years.
  • Available in small, trace-analysis pack sizes (e.g., 25 μg).