Learn More
Ampco 9/16 in. Tip Insulated Non-Sparking Wrench
SDP
Catalog No. 19174131 Shop All Ampco Metal Inc. Products
Change view
Click to view available options
Length (English):
6 to 1/4 in.
Catalog No. Length (English)
19-174-131 6 to 1/4 in.
1 options
Catalog No. 19-174-131 Supplier Ampco Supplier No. IWSB9/16
Add to Cart
Add to Cart
Insulated, non-magnetic, corrosion-resistant, and non-sparking wrench. Made in the USA.
  • Made in the USA.
  • HS Code: 8204.12
  • Length: 6 and 1/4 in.
Specifications
Length (English) 6 to 1/4 in.
Length (Metric) 158.75 mm
Material Aluminum Bronze
Product Type Insulated Wrench
WARNING: Cancer - www.P65Warnings.ca.gov