Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 9,19-Cyclolanostan-3-ol, 24-methylene-, 3-(4-hydroxy-3-methoxyphenyl)-2-propenoate, (3β)- | 469-36-3 | 5 MG
SDP

Catalog No. 5000417251
Encompass_Preferred

24-Methylenecycloartanyl ferulate is a γ-oryzanol compound. It promotes parvin-beta expression in human breast cancer cells and is a potential ATP-competitive Akt1 inhibitor.

  • Purity of 98.53%
  • White to off-white solid appearance
  • Soluble in DMSO at ≥ 12.5 mg/mL
  • Classified as a terpenoid and phenol
  • Derived from plants

Catalog No. 50-004-17251 Supplier Medchemexpress LLC Supplier No. HYN81225MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

24-Methylenecycloartanyl ferulate is a γ-oryzanol compound. It promotes parvin-beta expression in human breast cancer cells and is a potential ATP-competitive Akt1 inhibitor.

  • Purity of 98.53%
  • White to off-white solid appearance
  • Soluble in DMSO at ≥ 12.5 mg/mL
  • Classified as a terpenoid and phenol
  • Derived from plants

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.