Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 9-(2-deoxy-beta-D-ribofuranosyl)-6-methylpurine | 16006-64-7 | MFCD29059892 | 95.0% | 249.27 g·mol^-1 | C12H15N3O3 | 10 MG
SDP

Catalog No. 5000281312
Encompass_Preferred

9-(2-Deoxy-beta-D-ribofuranosyl)-6-methylpurine is a deoxyribonucleoside derivative of 6-methylpurine supplied as an off-white to light yellow powder for research use in biochemical and medicinal chemistry applications. The product is provided with analytical documentation and recommended cold-storage conditions.

  • Purity 95.0%.
  • CAS number 16006-64-7.
  • Molecular weight 249.27 g·mol^-1.
  • Supplied as an off-white to light yellow powder.
  • Recommended storage: powder at -20°C for long-term storage; in solvent at -80°C for extended stability.
  • Documentation available: data sheet, certificate of analysis, and safety data sheet.

Catalog No. 50-002-81312 Supplier Medchemexpress LLC Supplier No. HYW39443210MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

9-(2-Deoxy-beta-D-ribofuranosyl)-6-methylpurine is a deoxyribonucleoside derivative of 6-methylpurine supplied as an off-white to light yellow powder for research use in biochemical and medicinal chemistry applications. The product is provided with analytical documentation and recommended cold-storage conditions.

  • Purity 95.0%.
  • CAS number 16006-64-7.
  • Molecular weight 249.27 g·mol^-1.
  • Supplied as an off-white to light yellow powder.
  • Recommended storage: powder at -20°C for long-term storage; in solvent at -80°C for extended stability.
  • Documentation available: data sheet, certificate of analysis, and safety data sheet.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.