Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9-(2-Hydroxyethyl)anthracene 98.0+%, TCI America™
SDP

Catalog No. H09705G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H09705G 5 g
H09701G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H09705G Supplier TCI America Supplier No. H09705G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 54060-73-0
Molecular Formula C16H14O
Molecular Weight (g/mol) 222.287
MDL Number MFCD01318099
InChI Key KLNMQYHQWUWCPG-UHFFFAOYSA-N
Synonym 2-(9-Anthryl)ethanol
PubChem CID 281265
IUPAC Name 2-anthracen-9-ylethanol
SMILES C1=CC=C2C(=C1)C=C3C=CC=CC3=C2CCO

Specifications

Melting Point 125°C
Quantity 5 g
Formula Weight 222.29
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 9-(2-Hydroxyethyl)anthracene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.