Learn More
Ace Glass, Inc. 9.25 liter graduated cylindrical jar with side indents
SDP
Catalog No. 50972772
Encompass
9.25L graduated cylindrical jar with side indents for easier handling. Made of heavy wall glass. Also available with poly safety coating. Not for use with heat, pressure, or vacuum applications.
Catalog No. 50-972-772 Supplier Ace Glass, Inc. Supplier No. 623309
9.25L graduated cylindrical jar with side indents for easier handling. Made of heavy wall glass. Also available with poly safety coating. Not for use with heat, pressure, or vacuum applications.