Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9-[3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]carbazole 98.0+%, TCI America™
SDP

Catalog No. T33861G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T33861G 1 g
T33865G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T33861G Supplier TCI America Supplier No. T33861G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 870119-58-7
Molecular Formula C24H24BNO2
Molecular Weight (g/mol) 369.271
MDL Number MFCD22415352
InChI Key YWLGYAZJWNLVKB-UHFFFAOYSA-N
Synonym 3-(9H-Carbazol-9-yl)phenylboronic Acid Pinacol Ester, 2-[3-(9H-Carbazol-9-yl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
PubChem CID 58690899
IUPAC Name 9-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]carbazole
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC=C2)N3C4=CC=CC=C4C5=CC=CC=C53

Specifications

Melting Point 130°C
Color White
Quantity 1 g
Formula Weight 369.27
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 9-[3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]carbazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.