Learn More
Grainger 9.3 oz Aerosol Dry Lubricant
Catalog No. 502763749
Encompass_Preferred
The Blaster Multipurpose Grease is an aerosol lubricant designed for general lubrication and corrosion prevention. It contains lithium grease and is formulated to lubricate metal parts, hinges, slides, chains, cables, and more. The grease sprays on as a liquid and sets to a smooth, buttery grease. It adheres to metal and protects against rust and corrosion. The aerosol can contains 11 oz of grease and sprays up to 16 inches. It can be used in temperatures ranging from 0 to 300°F. The grease is NLGI Grade 2.
Catalog No. 50-276-3749 Supplier Grainger Supplier No. 16W547
The Blaster Multipurpose Grease is an aerosol lubricant designed for general lubrication and corrosion prevention. It contains lithium grease and is formulated to lubricate metal parts, hinges, slides, chains, cables, and more. The grease sprays on as a liquid and sets to a smooth, buttery grease. It adheres to metal and protects against rust and corrosion. The aerosol can contains 11 oz of grease and sprays up to 16 inches. It can be used in temperatures ranging from 0 to 300°F. The grease is NLGI Grade 2.