Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9-(4-Methoxyphenyl)xanthen-9-ol 97.0+%, TCI America™
SDP

Catalog No. M12255G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M12255G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M12255G Supplier TCI America Supplier No. M12255G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 94465-25-5
Molecular Formula C20H16O3
Molecular Weight (g/mol) 304.345
MDL Number MFCD00671561
InChI Key ZYACEJZTEMQQEU-UHFFFAOYSA-N
Synonym 9-(p-Anisyl)xanthen-9-ol, 9-Hydroxy-9-(4-methoxyphenyl)xanthene
PubChem CID 3439556
IUPAC Name 9-(4-methoxyphenyl)xanthen-9-ol
SMILES COC1=CC=C(C=C1)C2(C3=CC=CC=C3OC4=CC=CC=C42)O

Specifications

Melting Point 126°C
Quantity 5 g
Formula Weight 304.35
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 9-(4-Methoxyphenyl)xanthen-9-ol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.