Learn More
Electron Microscopy Sciences Cardboard 9/4 Microscope Slide Tray
SDP
Catalog No. 50931131
Encompass_Preferred
Heavy-duty cardboard construction with cloth hinged flaps to form a single compartment slide holder of various sizes horizontally. Holds 9 3x1" slides or 4 3x2" slides. The tray measures 235 x 78 mm (9 1/4" x 3 1/8")
Catalog No. 50-931-131 Supplier Electron Microscopy Sciences Supplier No. 7149712
Heavy-duty cardboard construction with cloth hinged flaps to form a single compartment slide holder of various sizes horizontally. Holds 9 3x1" slides or 4 3x2" slides. The tray measures 235 x 78 mm (9 1/4" x 3 1/8")