Learn More
Medchemexpress LLC 9-(8-azabicyclo[3.2.1]oct-3-ylidene)-9H-xanthene-3-carboxylic acid diethylamide | 825649-28-3 | 98.1% | 388.50 g/mol | C25H28N2O2 | 5 MG
SDP
Catalog No. 5000263604
Encompass_Preferred
JNJ-20788560 is a small-molecule research compound that acts as a selective and orally active delta opioid receptor (DOR) agonist, used in preclinical pharmacology studies of analgesia and inflammatory hyperalgesia.
  • Selective delta opioid receptor agonist with high affinity (≈2.0 nM).
  • Suitable for preclinical analgesia and inflammatory hyperalgesia research.
  • High purity solid (≈98.1%).
  • Molecular formula C25H28N2O2; molecular weight 388.50 g/mol.
  • Powder stable at -20°C for long-term storage; limited stability in solution.
Catalog No. 50-002-63604 Supplier Medchemexpress LLC Supplier No. HY110515MG
JNJ-20788560 is a small-molecule research compound that acts as a selective and orally active delta opioid receptor (DOR) agonist, used in preclinical pharmacology studies of analgesia and inflammatory hyperalgesia.
  • Selective delta opioid receptor agonist with high affinity (≈2.0 nM).
  • Suitable for preclinical analgesia and inflammatory hyperalgesia research.
  • High purity solid (≈98.1%).
  • Molecular formula C25H28N2O2; molecular weight 388.50 g/mol.
  • Powder stable at -20°C for long-term storage; limited stability in solution.