Learn More
Medchemexpress LLC [9,9'-Bianthracene]-2-carboxylic acid, 5,5'-bis(β-D-glucopyranosyloxy)-9,9',10,10'-tetrahydro... | 37271-16-2 | 848.76 | 5 MG
SDP
Catalog No. 5000415867
Encompass_Preferred
Sennoside C is an orally active component derived from the traditional Chinese medicine, Sennae Folium. It has the ability to bind to human lysozyme and inhibit its amyloid fibrosis with an IC50: 186.20 μM. The compound also demonstrates purgative activity and is suitable for research related to diarrhea and anti-amyloid related diseases.
  • Orally active component from the traditional Chinese medicine Sennae Folium.
  • Binds to human lysozyme and inhibits its amyloid fibrosis (IC50: 186.20 μM).
  • Possesses purgative activity.
  • Useful for research concerning diarrhea and anti-amyloid related diseases.
  • Demonstrated a purgative effect in mice via oral administration with an ED50 of 11.2 mg/kg.
  • Can enhance the purgative activity of Sennoside A when present in a mixture.
Catalog No. 50-004-15867 Supplier Medchemexpress LLC Supplier No. HYN19725MG
Sennoside C is an orally active component derived from the traditional Chinese medicine, Sennae Folium. It has the ability to bind to human lysozyme and inhibit its amyloid fibrosis with an IC50: 186.20 μM. The compound also demonstrates purgative activity and is suitable for research related to diarrhea and anti-amyloid related diseases.
  • Orally active component from the traditional Chinese medicine Sennae Folium.
  • Binds to human lysozyme and inhibits its amyloid fibrosis (IC50: 186.20 μM).
  • Possesses purgative activity.
  • Useful for research concerning diarrhea and anti-amyloid related diseases.
  • Demonstrated a purgative effect in mice via oral administration with an ED50 of 11.2 mg/kg.
  • Can enhance the purgative activity of Sennoside A when present in a mixture.