Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9,9-Bis(2-ethylhexyl)-2,7-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)fluorene 98.0+%, TCI America™
SDP

Catalog No. B48921G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B48921G 1 g
B4892200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B48921G Supplier TCI America Supplier No. B48921G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 357219-41-1
Molecular Formula C41H64B2O4
Molecular Weight (g/mol) 642.579
MDL Number MFCD16621099
InChI Key SMQUVDNJQVDOLI-UHFFFAOYSA-N
Synonym 9,9-Bis(2-ethylhexyl)fluorene-2,7-diboronic Acid Bis(pinacol) Ester
PubChem CID 12104084
IUPAC Name 2-[9,9-bis(2-ethylhexyl)-7-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)fluoren-2-yl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC3=C(C=C2)C4=C(C3(CC(CC)CCCC)CC(CC)CCCC)C=C(C=C4)B5OC(C(O5)(C)C)(C)C

Specifications

Melting Point 89°C
Color White-Yellow
Quantity 1 g
Formula Weight 642.58
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 9,9-Bis(2-ethylhexyl)-2,7-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)fluorene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.