Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9,9-Bis(4-bromophenyl)fluorene 98.0+%, TCI America™
SDP

Catalog No. B36455G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B36455G 5 g
B36451G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B36455G Supplier TCI America Supplier No. B36455G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 128406-10-0
Molecular Formula C25H16Br2
Molecular Weight (g/mol) 476.211
InChI Key VQTLUEKUYMRVDS-UHFFFAOYSA-N
PubChem CID 10695915
IUPAC Name 9,9-bis(4-bromophenyl)fluorene
SMILES C1=CC=C2C(=C1)C3=CC=CC=C3C2(C4=CC=C(C=C4)Br)C5=CC=C(C=C5)Br

Specifications

Melting Point 158°C
Color White
Quantity 5 g
Formula Weight 476.21
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 9,9-Bis(4-bromophenyl)fluorene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.