Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9,9-Bis(4-glycidyloxyphenyl)fluorene 98.0+%, TCI America™
SDP

Catalog No. B42695G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B42695G 5 g
B426925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B42695G Supplier TCI America Supplier No. B42695G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 47758-37-2
Molecular Formula C31H26O4
Molecular Weight (g/mol) 462.55
MDL Number MFCD02171148
InChI Key LCSAOPVSVLGDLE-UHFFFAOYNA-N
PubChem CID 93249
IUPAC Name 2-{[4-(9-{4-[(oxiran-2-yl)methoxy]phenyl}-9H-fluoren-9-yl)phenoxy]methyl}oxirane
SMILES C(OC1=CC=C(C=C1)C1(C2=CC=CC=C2C2=CC=CC=C12)C1=CC=C(OCC2CO2)C=C1)C1CO1

Specifications

Melting Point 159°C
Color White
Quantity 5 g
Synonym 4,4′C-(9-Fluorenylidene)bis(1,2-epoxy-3-phenoxypropane)
Formula Weight 462.55
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 9,9-Bis(4-glycidyloxyphenyl)fluorene
EINECSNumber (4)-1713
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.