Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9,9-Bis[4-[N-(1-naphthyl)anilino]phenyl]fluorene 97.0+%, TCI America™
SDP

Catalog No. B4979200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B4979200MG 200 mg
B49791G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B4979200MG Supplier TCI America Supplier No. B4979200MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 510775-24-3
Molecular Formula C57H40N2
Molecular Weight (g/mol) 752.961
InChI Key LKFCORSLOUZJMB-UHFFFAOYSA-N
PubChem CID 60106020
IUPAC Name N-[4-[9-[4-(N-naphthalen-1-ylanilino)phenyl]fluoren-9-yl]phenyl]-N-phenylnaphthalen-1-amine
SMILES C1=CC=C(C=C1)N(C2=CC=C(C=C2)C3(C4=CC=CC=C4C5=CC=CC=C53)C6=CC=C(C=C6)N(C7=CC=CC=C7)C8=CC=CC9=CC=CC=C98)C1=CC=CC2=CC=CC=C21

Specifications

Color White-Yellow
Quantity 200 mg
Formula Weight 752.96
Percent Purity ≥97.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 9,9-Bis[4-[N-(1-naphthyl)anilino]phenyl]fluorene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.