Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9,9-Dimethyl-10-phenyl-9,10-dihydroacridine 98.0+%, TCI America™
SDP

Catalog No. D5354200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D5354200MG 200 mg
D53541G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D5354200MG Supplier TCI America Supplier No. D5354200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 717880-39-2
Molecular Formula C21H19N
Molecular Weight (g/mol) 285.39
MDL Number MFCD01851187
InChI Key CKTUEZIFXPDRMO-UHFFFAOYSA-N
PubChem CID 66585389
IUPAC Name 9,9-dimethyl-10-phenylacridine
SMILES CC1(C2=CC=CC=C2N(C3=CC=CC=C31)C4=CC=CC=C4)C

Specifications

Melting Point 119°C
Color White-Yellow
Quantity 200 mg
Formula Weight 285.39
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 9,9-Dimethyl-10-phenyl-9,10-dihydroacridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.