Learn More
Strem, An Ascensus Company CAS# 856405-77-1. 500mg. 9,9-Dimethyl-4,5-bis(di-t-butylphosphino)xanthene, min. 97% t-Bu-XANTPHOS. MFCD07781995
SDP
Catalog No. 5090110126
Encompass
CAS# 856405-77-1. 500mg. 9,9-Dimethyl-4,5-bis(di-t-butylphosphino)xanthene, min. 97% t-Bu-XANTPHOS. MFCD07781995. Molecular Weight: 498.66. Molecular Formula: C31H48OP2. Color/form: white to light yellow pwdr. Strem# 15-1241. http://www.strem.com/catalog/v/15-1241/
Catalog No. 50-901-10126 Supplier Strem, An Ascensus Company Supplier No. 151241500MG
Add to Cart
Add to Cart
CAS# 856405-77-1. 500mg. 9,9-Dimethyl-4,5-bis(di-t-butylphosphino)xanthene, min. 97% t-Bu-XANTPHOS. MFCD07781995. Molecular Weight: 498.66. Molecular Formula: C31H48OP2. Color/form: white to light yellow pwdr. Strem# 15-1241. http://www.strem.com/catalog/v/15-1241/