Learn More
Medchemexpress LLC 9,9-dimethylfluoren-2-amine-d7 | 2837147-38-1 | 98.1% | 216.33 g·mol-1 | C15H8D7N | 250 MG
SDP
Catalog No. 5000302461
Encompass_Preferred
9,9-Dimethylfluoren-2-amine-d7 is the deuterium-labeled isotopologue of 9,9-dimethylfluoren-2-amine supplied as a high-purity solid for use in analytical and research applications. The deuterium enrichment provides a clear mass shift for reliable quantitation and trace-level detection in mass spectrometry and NMR workflows.
  • high-purity solid suitable for analytical use.
  • deuterium labeling enables unambiguous mass differentiation from native analytes.
  • compatible with NMR, GC-MS, and LC-MS workflows.
  • stable tracer for quantitative and method development studies.
Catalog No. 50-003-02461 Supplier Medchemexpress LLC Supplier No. 250MGHYW019610S1
Add to Cart
Add to Cart
9,9-Dimethylfluoren-2-amine-d7 is the deuterium-labeled isotopologue of 9,9-dimethylfluoren-2-amine supplied as a high-purity solid for use in analytical and research applications. The deuterium enrichment provides a clear mass shift for reliable quantitation and trace-level detection in mass spectrometry and NMR workflows.
  • high-purity solid suitable for analytical use.
  • deuterium labeling enables unambiguous mass differentiation from native analytes.
  • compatible with NMR, GC-MS, and LC-MS workflows.
  • stable tracer for quantitative and method development studies.