Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9,9-Dimethylfluoren-2-boronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. D39745G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D3974-5G 5 g
D39741G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D3974-5G Supplier TCI America Supplier No. D39745G
Add to Cart
Add to Cart

Specifications

CAS 333432-28-3
Color White-Yellow
Molecular Formula C15H15BO2
MDL Number MFCD08704227
Quantity 5 g
Synonym 9,9-dimethyl-9 h-fluoren-2-yl boronic acid, 9,9-dimethyl-9 h-fluoren-2-ylboronic acid, 9,9-dimethyl-9 h-fluoren-2-yl-boronic acid, 9,9-dimethylfluoren-2-boronic acid, 9,9-dimethyl-2-fluoreneboronic acid
InChI Key DMDPAJOXRYGXCB-UHFFFAOYSA-N
SMILES B(C1=CC2=C(C=C1)C3=CC=CC=C3C2(C)C)(O)O
IUPAC Name (9,9-dimethylfluoren-2-yl)boronic acid
Molecular Weight (g/mol) 238.093
PubChem CID 22348454
Formula Weight 238.09
Physical Form Crystalline Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.