Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9,9-Diphenylfluorene-2-boronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. D5084200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D5084200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D5084200MG Supplier TCI America Supplier No. D5084200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 400607-31-0
Color White
Molecular Formula C25H19BO2
Quantity 200 mg
MDL Number MFCD22581299
InChI Key CPFALCJMNUHBHP-UHFFFAOYSA-N
SMILES B(C1=CC2=C(C=C1)C3=CC=CC=C3C2(C4=CC=CC=C4)C5=CC=CC=C5)(O)O
IUPAC Name (9,9-diphenylfluoren-2-yl)boronic acid
Molecular Weight (g/mol) 362.235
PubChem CID 59343999
Formula Weight 362.24
Physical Form Crystalline Powder
Chemical Name or Material 9,9-Diphenylfluorene-2-boronic Acid (contains varying amounts of Anhydride)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.