Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9,9'-Spirobi[9H-fluorene]-2-boronic Acid, TCI America™
SDP

Catalog No. S08315G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S0831-5G 5 g
S08311G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S0831-5G Supplier TCI America Supplier No. S08315G
Add to Cart
Add to Cart

Specifications

Chemical Name or Material 9,9 '-Spirobi[9 H-fluorene]-2-boronic Acid
CAS 236389-21-2
Color White-Yellow
Molecular Formula C25H17BO2
Quantity 5 g
Synonym 9,9 '-spirobi fluoren-2-ylboronic acid, 9,9 '-spirobi 9 h-fluorene-2-boronic acid, b-9,9 '-spirobi 9 h-fluoren-2 '-yl-boronic acid, 9,9 '-spirobi fluorene-2-ylboronic acid, boronic acid, b-9,9 '-spirobi 9 h-fluoren-2 '-yl
InChI Key WDDLHUWVLROJLA-UHFFFAOYSA-N
SMILES B(C1=CC2=C(C=C1)C3=CC=CC=C3C24C5=CC=CC=C5C6=CC=CC=C46)(O)O
IUPAC Name 9,9 '-spirobi[fluorene]-2-ylboronic acid
Molecular Weight (g/mol) 360.219
PubChem CID 22564369
Formula Weight 360.22
Physical Form Crystalline Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.