Learn More
Apexbio Technology LLC 9-AC 723-62-6 100mg
SDP
Catalog No. 502240720
Encompass_Preferred
9-AC (CAS 723-62-6) also known as anthracene-9-carboxylic acid is an anthracene derivative commonly utilized as an inhibitor of Ca -activated Cl channels (CaCCs) It selectively blocks Ca -dependent chloride conductance in various cell types including smooth muscle epithelial and salivary gland cells By modulating ion channel activity 9-AC serves as a valuable tool for investigating cellular electrophysiological processes signal transduction pathways and ion channel pharmacology Its inhibitory action on CaCCs enables studies of cell excitability regulation and chloride transport in both physiological and pathological contexts
This item is not returnable. View return policy
Catalog No. 50-224-0720 Supplier Apexbio Technology LLC Supplier No. B6525100
9-AC (CAS 723-62-6) also known as anthracene-9-carboxylic acid is an anthracene derivative commonly utilized as an inhibitor of Ca -activated Cl channels (CaCCs) It selectively blocks Ca -dependent chloride conductance in various cell types including smooth muscle epithelial and salivary gland cells By modulating ion channel activity 9-AC serves as a valuable tool for investigating cellular electrophysiological processes signal transduction pathways and ion channel pharmacology Its inhibitory action on CaCCs enables studies of cell excitability regulation and chloride transport in both physiological and pathological contexts