Learn More
Medchemexpress LLC 9-acetyl-3,4-dihydro-2H-carbazol-1-one | 352553-42-5 | 99.8% | 227.26 g/mol | C14H13NO2 | 10 MG
SDP
Catalog No. 5000229749
Encompass_Preferred
MS7972 is a small-molecule inhibitor that disrupts the interaction between human p53 and CREB binding protein (CBP). It has been reported to nearly completely block this bromodomain (BRD) interaction at 50 μM and is supplied as a purified research compound for biochemical and cellular studies.
  • Blocks p53-CREB binding nearly completely at 50 μM.
  • Suitable for biochemical and cell-based assays.
  • High purity (99.81%) and characterized by NMR and HPLC.
  • Well-defined small-molecule structure: C14H13NO2, MW 227.26 g/mol.
  • Available in small-scale quantities for research use, including 10 MG packages.
Catalog No. 50-002-29749 Supplier Medchemexpress LLC Supplier No. HY11905310MG
MS7972 is a small-molecule inhibitor that disrupts the interaction between human p53 and CREB binding protein (CBP). It has been reported to nearly completely block this bromodomain (BRD) interaction at 50 μM and is supplied as a purified research compound for biochemical and cellular studies.
  • Blocks p53-CREB binding nearly completely at 50 μM.
  • Suitable for biochemical and cell-based assays.
  • High purity (99.81%) and characterized by NMR and HPLC.
  • Well-defined small-molecule structure: C14H13NO2, MW 227.26 g/mol.
  • Available in small-scale quantities for research use, including 10 MG packages.