Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9-Acetylanthracene, 95%

Catalog No. AC102440250
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC102440250 25 g Glass bottle
AC102440050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC102440250 Supplier Thermo Scientific Chemicals Supplier No. 102440250
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 784-04-3 | C16H12O | 220.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 784-04-3
Molecular Formula C16H12O
Molecular Weight (g/mol) 220.27
MDL Number MFCD00001259
InChI Key NXXNVJDXUHMAHU-UHFFFAOYSA-N
Synonym 9-acetylanthracene, 1-anthracen-9-yl ethanone, 9-acetoanthracene, 9-anthryl methyl ketone, ethanone, 1-9-anthracenyl, 1-9-anthryl ethanone, ketone, 9-anthryl methyl, 9-acetyl anthracene, 9-acetylanthrancene, 9-acetylantracene
PubChem CID 69911
IUPAC Name 1-anthracen-9-ylethanone
SMILES CC(=O)C1=C2C=CC=CC2=CC3=CC=CC=C31

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 72°C to 76°C
Color Brown-Yellow to Yellow
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Quantity 25 g
Beilstein 07, II, 450
Formula Weight 220.27
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material 9-Acetylanthracene, 95%
Show More Show Less
EINECSNumber 212-315-2

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.