Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9-Acridinecarboxaldehyde, 97%, Thermo Scientific Chemicals

Catalog No. AC452050050
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC452050050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC452050050 Supplier Thermo Scientific Chemicals Supplier No. 452050050

missing translation for 'validMainframeMsg'

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 885-23-4
Molecular Formula C14H9NO
Molecular Weight (g/mol) 207.23
InChI Key ISOCABSXIKQOOV-UHFFFAOYSA-N
Synonym 9-formylacridine, 9-acridinecarboxaldehyde, 9-acridinealdehyde, acridine, 9-formyl, acridine-9-carboxaldehyde, 9-acridinecarboxaldehyde 8ci 9ci, 9-acridine carboxaldehyde, 9-acridinecarbaldehyde
PubChem CID 98663
IUPAC Name acridine-9-carbaldehyde
SMILES C1=CC=C2C(=C1)C(=C3C=CC=CC3=N2)C=O

Specifications

Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Quantity 5 g
Formula Weight 207.23
Percent Purity 97%
Chemical Name or Material 9-Acridinecarboxaldehyde
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.